Mix & Go E. coli Transformation Kit and Buffer Set
Mix & Go E. coli transformation kit, 1 KIT
Katalognr. ZYMOT3001
Marketsource
Anbieter: Zymo Research
Mix & Go E. coli Transformation Kit and Buffer Set
Katalognr. ZYMOT3001
Anbieter: Zymo Research
Marketsource
CAS Nummer:
Direkt zu
Product Details & Documents
The Mix & Go kit convenient for the preparation of competent E. coli.
- It is very simple and efficient DNA transformation
- It completely eliminates the requirement for heat shocking and related procedures
- Transformation efficiencies are typically on the order of 108 to 109 transformants/µg plasmid DNA
The Mix & Go E. coli transformation kit includes all buffers and ZymoBroth™ medium to generate 20 ml of Mix & Go competent cells.
Technische Daten
Technische Daten
- VE:1 KIT
- Bezeichnung:Mix & Go E. coli transformation kit
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.