DNase I
1 set, 1 KIT
ZYMOE1010
Marketsource
DNase I (RNase-free) cuts both double-stranded and single-stranded DNA.
- Lyophilized enzyme
- Used for selectively degrading DNA in the presence of RNA
DNase is suited for applications such as nick translation, production of random fragments, cleavage of genomic DNA for footprinting, removal of DNA template after in vitro transcription and removal of DNA from RNA samples prior to applications such as RT-PCR. It is compatible with all of our RNA kits featuring in-column DNase digestion. Size 250 U.
: The set supplied with DNA digestion buffer (4 ml).
- VE:1 KIT
- Pk Info:1 set
- Cat. No.:ZYMOE1010
- Enzymname:DNase I