Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Proteinase K | Avantor

Proteinase K

Marketsource
Anbieter:   Zymo Research
Proteinase K
Anbieter:  Zymo Research
Marketsource
Zertifikate
Proteinase K

2 Varianten

Product Details & Documents

Proteinase K is a stable serine protease with broad substrate specificity and will degrade many proteins in their native conformation, even in the presence of detergents (e.g., SDS).

  • Specifications: Lyophilized enzyme provided with Proteinase K Storage Buffer.
  • Enzyme Commission Number: (EC 3.4.21.64)
  • Source: Engyodontium album pH and Temperature Range: 4,0 to 12,0 (8,0 is optimum), 25 to 65 °C.
  • Specific Activity: Over 30 units/mg protein
  • Unit Definition: One unit (U) of enzyme will hydrolyze urea-denatured hemoglobin to produce 1.0 μmole of tyrosine per minute at pH 7,5 at 37 °C.

The enzyme is frequently used in molecular biology applications to digest unwanted proteins such as nucleases from DNA and/or RNA preparations from microorganisms, cells, and plants.

Spezifikationen für diese Produktfamilie

Technische Daten

Technische Daten

  • Catalog No:
  • ZYMOD3001-2-5
  • ZYMOD3001-2-20
  • Enzymname:
  • Proteinase K
  • Proteinase K

Technische Daten

Product Family Options

Produktinformationen

  • Pk Info
    VE
    Verfügbarkeit
    Preis
  • 1×5 mg
    1 SET
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 SET
    1 * 1 SET - SFr. 31.00
    SFr. 31.00
    1 * 1 SET
       SFr. 31.00

    Spezifikationen:
    Marketsource
    Mehr
    Specifications
    Cat. No.
    ZYMOD3001-2-5
    Enzymname
    Proteinase K
    Marketsource
  • 1×20 mg
    1 SET
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 SET
    1 * 1 SET - SFr. 64.00
    SFr. 64.00
    1 * 1 SET
       SFr. 64.00

    Spezifikationen:
    Marketsource
    Mehr
    Specifications
    Cat. No.
    ZYMOD3001-2-20
    Enzymname
    Proteinase K
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.