96-Well Blocks
Marketsource
Supplier: Zymo Research
Jump to:
About this item
Deep-well block, 2 ml for processing and mixing of samples.
- Conforms to laboratory standards
96-well block features durable, clear polypropylene construction. Each has a level footprint and conforms to laboratory standards. Capacity is 2 ml per round bottom well.
3 Options Available Below
Product Details & Documents
Deep-well block, 2 ml for processing and mixing of samples.
- Conforms to laboratory standards
96-well block features durable, clear polypropylene construction. Each has a level footprint and conforms to laboratory standards. Capacity is 2 ml per round bottom well.
Product Family Options
Product Information
- BezeichnungVerpacktGesamtvolumenAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource