Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Zymo-Spin™ IC Columns
Marketsource
Zymo-Spin™ IC Columns
Marketsource
Zymo-Spin™ IC Columns
Zymo-Spin™ IC Columns
Spin columns for the purification of DNA or RNA.
  • Format: DNA/RNA binding
  • DNA binding capacity: 5 µg
  • RNA binding capacity: 10 µg
  • Compatibility: microcentrifuge, vacuum manifold
Capped version of the Zymo-Spin™ I column. The Zymo-Spin™ IC column can be used either in microcentrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin™ IC features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 μg of DNA, and 10 μg of RNA, in ≥ 6 μl eluate. Capacity is 800 μl.
Product Family Options
  • Bezeichnung
    Format
    Packung
    VE
  • Zymo-Spin™ IC
    Spin Column
    250 pack
    250
    1 * 250 ST
    1 * 250 ST - SFr. 328.00
    SFr. 328.00
    1 * 250 ST
       SFr. 328.00
    Marketsource
    Marketsource
  • Zymo-Spin™ IC
    Spin Column
    50 pack
    50
    1 * 50 ST
    1 * 50 ST - SFr. 77.00
    SFr. 77.00
    1 * 50 ST
       SFr. 77.00
    Marketsource
    Marketsource