Hexythiazox Analytical standard, reference material, Fluka™
Hexythiazox, Analytical standard, Glass Bottle, 100 mg
HONCR1983-100MG
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Packung:Glass Bottle
- VE:100 mg
- Normen:Analytical standard
- Bezeichnung:Hexythiazox
- Cat. No.:HONCR1983-100MG
- CAS Nr.:78587-05-0
- Molekulargewicht:352.88 g/mol
- Physical form:Solid
- MDL No.:MFCD00137385
- Synonym(s):(4RS,5RS)-5-(4-Chlorophenyl)-N-cyclohexyl-4-methyl-2-oxothiazolidine-3-carboxamide
- Bezeichnung:Hexythiazox
- Schmelzpunkt:106 °C
- Formel:C₁₇H₂₁ClN₂O₂S



