Chloramphenicol Analytical standard, reference material, Fluka™
Chloramphenicol, Analytical standard, Glass Bottle, 100 mg
Katalognr. HONCR1900-100MG
Anbieter: Honeywell Chemicals
Chloramphenicol Analytical standard, reference material, Fluka™
Katalognr. HONCR1900-100MG
Anbieter: Honeywell Chemicals
CAS Nummer:
Direkt zu
Product Details & Documents
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Technische Daten
Technische Daten
- Packung:Glass Bottle
- VE:100 mg
- Normen:Analytical standard
- Bezeichnung:Chloramphenicol
- Cat. No.:HONCR1900-100MG
- CAS Nr.:56-75-7
- Molekulargewicht:323.13 g/mol
- Physical form:Solid
- MDL No.:MFCD00078159
- Lagerbedingungen:RT
- Synonym(s):D-(-)-threo-2,2-Dichloro-N-[ß-hydroxy-a-(hydroxymethyl)-ß-(4-nitrophenyl)ethyl]acetamide
- Bezeichnung:Chloramphenicol
- Schmelzpunkt:151 °C
- Formel:C₁₁H₁₂Cl₂N₂O₅



