Carfentrazone-ethyl Analytical standard, reference material, Fluka™
Carfentrazone-ethyl, Analytical standard, Glass Bottle, 25 mg
Codice catalogo HONCR1844-25MG
Nome del fornitore: Honeywell Chemicals
Carfentrazone-ethyl Analytical standard, reference material, Fluka™
Codice catalogo HONCR1844-25MG
Nome del fornitore: Honeywell Chemicals
Numero CAS:
Vai a
Product Details & Documents
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifiche
Specifiche
- Packung:Glass Bottle
- VE:25 mg
- Normen:Analytical standard
- Bezeichnung:Carfentrazone-ethyl
- Cat. No.:HONCR1844-25MG
- CAS Nr.:128639-02-1
- Molekulargewicht:412.19 g/mol
- Flammpunkt:229 °C
- Physical form:Liquid
- MDL No.:MFCD03792738
- Lagerbedingungen:2-8°C
- Synonym(s):Ethyl 2-chloro-3-{2-chloro-5-[4-(difluoromethyl)-4,5-dihydro-3-methyl-5-oxo-1H-1,2,4-triazol-1-yl]-4-fluoro-phenyl}propionate
- Bezeichnung:Carfentrazone-ethyl
- Schmelzpunkt:-22 °C
- Formel:C₁₅H₁₄Cl₂F₃N₃O₃



