Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Oryzalin Analytical standard, reference material, Fluka™

Oryzalin Analytical standard, reference material, Fluka™

Oryzalin, Analytical standard, Glass Bottle, 100 mg
Catalog # HONCR1789-100MG
Supplier:   Honeywell Chemicals
Oryzalin Analytical standard, reference material, Fluka™
Catalog # HONCR1789-100MG
CAS Number:  
Oryzalin Analytical standard, reference material, Fluka™

Oryzalin, Analytical standard, Glass Bottle, 100 mg

CAS Number:  
Price
SFr. 66.98
Log in for availability
1 * 100 mg
Product Details & Documents

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
Specifications

Specifications

  • Packung:
    Glass Bottle
  • VE:
    100 mg
  • Normen:
    Analytical standard
  • Bezeichnung:
    Oryzalin
  • Cat. No.:
    HONCR1789-100MG
  • CAS Nr.:
    19044-88-3
  • Molekulargewicht:
    346.36 g/mol
  • Physical form:
    Crystalline
  • MDL No.:
    MFCD00078725
  • Lagerbedingungen:
    RT
  • Synonym(s):
    4-(dipropylamino)-3,5-dinitrobenzenesulfonamide
  • Bezeichnung:
    Oryzalin
  • Schmelzpunkt:
    139.7 °C
  • Formel:
    C₁₂H₁₈N₄O₆S

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.