AuftragsverfolgungKontakt
Gadoteridol, Reference Standards
Gadoteridol, Reference Standards
Supplier:   USP
Gadoteridol, Reference Standards
Supplier:  USP
Certificates
4 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 1287631.
  • 1287642.
  • 1287653.
  • 1287664.
  • Bezeichnung:
  • Gadoteridol
  • Gadoteridol Related Compound A
  • Gadoteridol Related Compound B
  • Gadoteridol Related Compound C
  • Synonym(s):
  • 10-(2-hydroxypropyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triacetic acid
  • 1,4,7,10-tetraazacyclododecane-1,4,7-triacetic acid gadolinium salt
  • 1,4,7,10-Tetraaza-11-oxo-bicyclo[8.2.2]tetradecane-4,7-diacetic acid
  • CAS Nr.:
  • 120066-54-8
  • 120041-08-9
  • 112188-16-6
  • 220182-19-4
Specifications
Product Family Options
Product Information
  • Bezeichnung
    Normen
    Packung
    VE
    Availability
    Price
  • Gadoteridol
    USP reference standard
    Vial
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - SFr. 527.00
    SFr. 527.00
    1 * 500 mg
       SFr. 527.00
    Specifications:
    More
    Specifications
    Cat. No.
    1287631.
    Bezeichnung
    Gadoteridol
    Synonym(s)
    CAS Nr.
    120066-54-8
  • Gadoteridol Related Compound A
    USP reference standard
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - SFr. 1’370.00
    SFr. 1’370.00
    1 * 50 mg
       SFr. 1’370.00
    Specifications:
    More
    Specifications
    Cat. No.
    1287642.
    Bezeichnung
    Gadoteridol Related Compound A
    Synonym(s)
    10-(2-hydroxypropyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triacetic acid
    CAS Nr.
    120041-08-9
  • Gadoteridol Related Compound B
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - SFr. 1’440.00
    SFr. 1’440.00
    1 * 50 mg
       SFr. 1’440.00
    Specifications:
    More
    Specifications
    Cat. No.
    1287653.
    Bezeichnung
    Gadoteridol Related Compound B
    Synonym(s)
    1,4,7,10-tetraazacyclododecane-1,4,7-triacetic acid gadolinium salt
    CAS Nr.
    112188-16-6
  • Gadoteridol Related Compound C
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - SFr. 1’290.00
    SFr. 1’290.00
    1 * 50 mg
       SFr. 1’290.00
    Specifications:
    More
    Specifications
    Cat. No.
    1287664.
    Bezeichnung
    Gadoteridol Related Compound C
    Synonym(s)
    1,4,7,10-Tetraaza-11-oxo-bicyclo[8.2.2]tetradecane-4,7-diacetic acid
    CAS Nr.
    220182-19-4
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..