Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

4-Methylacetophenone, Pharmaceutical Analytical Impurities

4-Methylacetophenone, Pharmaceutical Analytical Impurities, Ampule, 25 mg
  1A00900.
 :   USP
4-Methylacetophenone, Pharmaceutical Analytical Impurities
  1A00900.
 :  USP
 :  
4-Methylacetophenone, Pharmaceutical Analytical Impurities

4-Methylacetophenone, Pharmaceutical Analytical Impurities, Ampule, 25 mg

 :  
 
SFr. 700.00
   
1 * 25 mg
 

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.

 

 

  • Packung:
    Ampule
  • VE:
    25 mg
  • Normen:
    Pharmaceutical Analytical Impurities
  • Bezeichnung:
    4-Methylacetophenone
  • Cat. No.:
    1A00900.
  • CAS Nr.:
    122-00-9
  • Synonym(s):
    1-(4-Methylphenyl)-1-ethanone (4'-Methylacetophenone)
  • Bezeichnung:
    4-Methylacetophenone