AuftragsverfolgungKontakt
Sash-glider kit | Avantor
Sash-glider kit
Sash-glider kit
Katalognr. LBCP3408500
Anbieter:   LABCONCO
Sash-glider kit
Katalognr. LBCP3408500
Anbieter:  LABCONCO
Sash-glider kit
Sash-glider kit
Sash-glider kit
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 KIT
Product Details & Documents
Use the Sash-glider to clean both sides of a biological safety cabinet's sash while it remains closed, to avoid contamination risks.
 : Sash-glider kit includes magnetic outside and inside blades, 2 microfibre sheets and 10 disposable white scrubbing strips. Additional scrubbing strips are available.
Technische Daten
  • Bezeichnung:
    Sash-glider kit