SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Zentrifugen
Rotoren
Swinging-bucket rotor package, SW 55 Ti, Optima
Swinging-bucket rotor package, SW 55 Ti, Optima
521-3731
:
Beckman Coulter
Swinging-bucket rotor package, SW 55 Ti, Optima
521-3731
:
Beckman Coulter
Bezeichnung:
SW 55 Ti swinging bucket rotor package
VE:
1
Für:
Swinging bucket (ultracentrifugation)
Max. RPM:
55000
No. of tubes accepted:
6×5 ml
Radius:
108,5 mm