ContactUsLinkComponent
Dumont #5AC anti-capillary forceps
Catalog # FSCI11258-20
Supplier:  FINE SCIENCE TOOLS
undefined
Dumont #5AC anti-capillary forceps
Catalog # FSCI11258-20
Supplier:  FINE SCIENCE TOOLS
Specifications
  • Typ:
    #5 anti-capillary
  • Länge:
    115 mm
  • Material:
    Inox
  • Spitzentyp:
    Biologie
  • VE:
    1
  • Spitzengröße:
    0,07×0,02 mm
  • Spitzenform:
    Gerade