Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pinzetten
Student Dumont #5 forceps
Student Dumont #5 forceps
Student Dumont #5 forceps
Catalog # FSCI91150-20
Supplier:
FINE SCIENCE TOOLS
Expand image
Student Dumont #5 forceps
Catalog # FSCI91150-20
Supplier:
FINE SCIENCE TOOLS
Specifications
Typ:
#5
Länge:
110 mm
Material:
Inox
Spitzentyp:
Standard
VE:
1
Spitzengröße:
0,1×0,06 mm
Spitzenform:
Gerade
Specifications
About this item
Product Details
Product Details