Titanium forceps
Titan, 160 mm, Serrated, Gerade, 0,3×0,5 mm
Catalog # FSCI11602-16
Marketsource
Supplier: FINE SCIENCE TOOLS
Jump to:
Product Details & Documents
Specifications
- Länge:160 mm
- Material:Titan
- Spitzentyp:Serrated
- Spitzengröße:0,3×0,5 mm
- Spitzenform:Gerade