Fine scissors, sharply angled up
Catalog # FSCI14037-10
Supplier: FINE SCIENCE TOOLS
Specifications
- Typ:Sharp/sharp, angled up tips
- Länge:105 mm
- VE:1
- Spitzenlänge:14 mm
Specifications
About this item
These scissors are made of stainless steel.