Micro volume Pipette tips, MµltiFit™
Supplier: Sorenson Bioscience
Certificates
About this item
These natural-colored tips fit most popular brand research-grade pipettors.
They feature a narrow bore that reduces dead air space around extended air-displacement piston. Narrow bore also increases accuracy and reproducibility for critical ultra microvolume ranges.
Certifications: Certified RNase-, DNase-, human DNA-, PCR inhibitor-, and pyrogen-free.
Ordering information: Available in autoclavable polypropylene racks, bulk packed, or QuickRack reloads for the reloading racks.
Packaging: Contents: Casein peptone (10 g/L), UF Yeast Powder (5 g/L), NaCl (10 g/L)
Frequently Bought Together