Order Entry
Switzerland
ContactUsLinkComponent
Anti-STAT3 (phospho-Tyr705) Mouse Monoclonal Antibody [clone: 13A3-1] (Brilliant Violet® 421)
Anbieter:  Biolegend
Zertifikate
Anti-STAT3 (phospho-Tyr705) Mouse Monoclonal Antibody [clone: 13A3-1] (Brilliant Violet® 421)
Anbieter:  Biolegend

Technische Daten

  • Catalog No:
  • BLEG651010
  • BLEG651009
  • Antigen-Symbol:
  • STAT3
  • STAT3
  • Antigen-Synonyme:
  • Signal transducer and activator of transcription 3|Acute-phase response factor|HIES|APRF
  • Signal transducer and activator of transcription 3|Acute-phase response factor|HIES|APRF
  • Klonalität:
  • Monoklonal
  • Monoklonal
  • Konjugation:
  • Brilliant Violet® 421
  • Brilliant Violet® 421
  • Klon:
  • 13A3-1
  • 13A3-1
  • Reaktivität:
  • Human, Mouse
  • Human, Mouse
  • Wirt:
  • Mouse
  • Mouse
  • Isotyp:
  • IgG1 kappa
  • IgG1 kappa
  • Durchflusszytometrie:
  • Yes
  • Yes
  • Modification:
  • phospho-Tyr705
  • phospho-Tyr705
  • Immunogen:
  • KLH conjugated modified synthetic peptide
  • KLH conjugated modified synthetic peptide
  • Reinigung:
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions.
  • The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions.
  • Lagerungspuffer:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
  • Lagertemperatur:
  • 2…8 °C
  • 2…8 °C
  • Validierte Anwendungen:
  • ICFC
  • ICFC

Technische Daten

Frequently Bought Together