Anti-CD279 (PD-1) Mouse Monoclonal Antibody [clone: EH12.2H7] (Brilliant Violet® 785)
Catalog # BLEG329930
Supplier: Biolegend
Specifications
- Antigen-Symbol:CD279 (PD-1)
- Klonalität:Monoklonal
- Klon:EH12.2H7
- Konjugation:Brilliant Violet® 785
- Durchflusszytometrie:Yes
- Wirt:Mouse
- Isotyp:IgG1 kappa
- Reaktivität:Baboon,African green monkey,Cynomolgus monkey,Common marmoset,Rhesus monkey,Chimpanzee,Human,Squirrel monkey
- Antigen-Synonyme:PD-1
- Lagerungspuffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Lagertemperatur:2…8 °C
- Validierte Anwendungen:FC
- Reinigung:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 785™ under optimal conditions.
- Größe:100 tests
- VE:100 Tests