Cone supplies
Product Details & Documents
A range of accessories and supplies for skimmer and sampling cones for use with our ICP-MS portfolio.
- Viton O-ring for sampling cone that seals the sampling cone to the vacuum chamber
- Screws for skimmer bases are available and used to install the sampling base to the vacuum chamber
- They also provide round interface wrenches (in order to remove and install the skimmer cone) and sampling cone wrenches (used to remove and install the sampling cone)
High quality ICP-MS systems contain interfaces that comprise of a step-down vacuum stage, located between a pair of conical metal plates, known as interface cones. In common terminology, the first and second cones are referred to as the sample cone and skimmer cone, respectively. A range of accessories, components and spare parts are available to enhance the performance of interface cones and for replacement purposes. To ensure system accuracy and longevity, replace cone supplies in accordance with recommended maintenance schedules.
Skimmer base (with screws) in stainless steel or brass option. The stainless steel base holds the standard nickel skimmer cone - do not use with platinum cone. The brass base can hold the platinum or nickel skimmer cone.
Specifications
- Für:ICP-MS portfolio
- Bezeichnung:ICP-MS detergent, for cleaning sampling cones, skimmer and lenses, 1 gallon