SIPS 10/20 pump tubing
VARI9910076100
: VARIAN
- Bezeichnung:Tubing kit for SIPS 10/20, to connect constant pressure head bottle to tee piece
- VE:1
SIPS 10/20 pump tubing for atomic absorption spectroscopy.
- Selected to provide extended lifetime and accurate dilution when using the SIPS
The Sample Introduction Pump System (SIPS) is an innovative sample introduction and dilution system for flame AA that eliminates the task of manually preparing multiple calibration standards, providing fast, accurate on-line dilution of over range samples.