Burner system components for PerkinElmer
Über diesen Artikel
Agilent burner components for PerkinElmer AA systems are designed to provide the best precision, efficient drainage, minimal burner blockage, and reduced interferences.
- Guaranteed to be compatible with PerkinElmer instruments
- Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies.
19 Varianten
Product Details & Documents
Agilent burner components for PerkinElmer AA systems are designed to provide the best precision, efficient drainage, minimal burner blockage, and reduced interferences.
- Guaranteed to be compatible with PerkinElmer instruments
- Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies.
Product Family Options
Produktinformationen
- BezeichnungVerfügbarkeitPreis
- Burner drain assembly for organics, for AAnalyst 100/200/300/400/700/800Einloggen für Verfügbarkeit1 * 1 ST
- Burner drain assembly, with tubing and drain interlock, drain bottle not included, for AAnalyst 100/200/300/400/700/800Einloggen für Verfügbarkeit1 * 1 ST
- Burner drain assembly, with tubing and drain interlock, drain bottle not included, for PinAAcleEinloggen für Verfügbarkeit1 * 1 ST
- Burner gasket kit for end cap assembly metal body nebulisers, for AAnalyst 100/200/300/400/700/800Einloggen für Verfügbarkeit1 * 1 ST
- Clamping ring for burner system, for AAnalyst 100/200/300/400/700/800Einloggen für Verfügbarkeit1 * 1 ST
- Drain tubing, tygon, with FEP lining, for AAnalyst 100/200/300/400/700/800Einloggen für Verfügbarkeit1 * 1 ST
- End cap assembly, for plastic body nebulizers, for AAnalyst 100/200/300/400/700/800Einloggen für Verfügbarkeit1 * 1 ST
- Gasket (O-ring), corkpreen, for burner end cap organics, for AAnalyst 100/200/300/400/700/800Einloggen für Verfügbarkeit1 * 1 ST
- O-ring kit, viton, for burner head, for AAnalyst 100/200/300/400/700/800Einloggen für Verfügbarkeit1 * 1 ST
- O-ring, ¹/₄", Int.Ø 6 mm, for end cap assembly for metal body nebulizers, for AAnalyst 100/200/300/400/700/800Einloggen für Verfügbarkeit1 * 1 ST
- O-ring, ¹/₈", Int.Ø 3 mm, for seal plug in end cap assembly, for metal body nebulisers, for PinAAcle and AAnalyst 100/200/300/400/700/800Einloggen für Verfügbarkeit1 * 1 ST
- Seal plug, pyrex, for impact bead used with end cap assembly, for AAnalyst 100/200/300/400/700/800Einloggen für Verfügbarkeit1 * 1 ST