Cap strips for Piko® UTW plates
Katalognr. 731-0392
Anbieter: Thermo Fisher Scientific
Technische Daten
- Bezeichnung:Streifen mit 8 durchsichtigen flachen Kappen für Piko® PCR-Platten mit ultradünnen Wänden
- Environmentally Preferable:
- VE:120
Technische Daten
Über diesen Artikel
These flat cap strips provide an extremely tight seal in UTW (ultra-thin walled) PCR plates. They are an ideal choice for low volume reactions where it is important to minimize vapor loss.
- 8-cap strip
- Optically clear for fluorescence detection
- Tight sealing for low-volume samples and consistent quantification results
- Ideal for sealing 24-well Piko UTW PCR plates
The optically-clear flat caps provide great transmission for all fluorescence detection applications, such as qPCR.