Recombinant Tick-Borne Encephalitis Virus NE/gE
Catalog # APOSBITP1920-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- VE:1 mg
- Protein/Peptid-Typ:Rekombinant
- Proteinsynonym:Encephalitis NE/gE
- Protein/Peptid-Name:Tick-Borne Encephalitis Virus NE/gE