ContactUsLinkComponent
Recombinant HIV-1 nef
Catalog # APOSBITP1866-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant HIV-1 nef
Catalog # APOSBITP1866-1MG
Supplier:  Apollo Scientific
CAS Number:  
Specifications
  • VE:
    1 mg
  • Protein/Peptid-Typ:
    Rekombinant
  • Protein/Peptid-Name:
    HIV-1 nef