Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteine und Peptide
Recombinant Treponema pallidum p17
Recombinant Treponema pallidum p17
Recombinant Treponema pallidum p17
APOSBITP1857-1MG
:
Apollo Scientific
:
Recombinant Treponema pallidum p17
APOSBITP1857-1MG
:
Apollo Scientific
:
VE:
1 mg
Protein/Peptid-Typ:
Rekombinant
Proteinsynonym:
Treponema p17
Protein/Peptid-Name:
Treponema pallidum p17