Recombinant Hepatitis C Virus NS5 Genotype-3
Catalog # APOSBITP1801-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- VE:1 mg
- Protein/Peptid-Typ:Rekombinant
- Proteinsynonym:HCV NS5 Genotype-3
- Protein/Peptid-Name:Hepatitis C Virus NS5 Genotype-3