Recombinant Hepatitis C Virus NS3, HRP (Horseradish Peroxidase)
Catalog # APOSBITP1782-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- VE:1 mg
- Konjugation:HRP (Horseradish Peroxidase)
- Protein/Peptid-Typ:Rekombinant
- Proteinsynonym:HCV NS3
- Protein/Peptid-Name:Hepatitis C Virus NS3