SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Proteine und Peptide
Recombinant Hepatitis C Virus NS3 Genotype-3 (1356-1459)
Recombinant Hepatitis C Virus NS3 Genotype-3 (1356-1459)
APOSBITP1776-1MG
:
Apollo Scientific
:
Recombinant Hepatitis C Virus NS3 Genotype-3 (1356-1459)
APOSBITP1776-1MG
:
Apollo Scientific
:
VE:
1 mg
Protein/Peptid-Typ:
Rekombinant
Proteinsynonym:
HCV NS3 Genotype-3
Protein/Peptid-Name:
Hepatitis C Virus NS3 Genotype-3 (1356-1459)