Recombinant Hepatitis C Virus NS3 Genotype-1b (1192-1459)
Catalog # APOSBITP1765-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- VE:1 mg
- Protein/Peptid-Typ:Rekombinant
- Proteinsynonym:HCV NS3 Genotype-1b
- Protein/Peptid-Name:Hepatitis C Virus NS3 Genotype-1b (1192-1459)