Skip to Header
Skip to Main Content
Skip to Footer
Erweiterte Suche
Erweiterte Suche
Sicherheitsdatenblatt (SDS)
Zertifikate
Herstellerdaten
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Express Eingabe
Bestellstatus
Ihr Konto
Warenkorb
Switzerland
Express Eingabe
Bestellstatus
Ihr Konto
Warenkorb
ContactUsLinkComponent
Home
Proteine und Peptide
Human Recombinant STAT3
Human Recombinant STAT3
Human Recombinant STAT3
Catalog # APOSBITP1419-5UG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Human Recombinant STAT3
Catalog # APOSBITP1419-5UG
Supplier:
Apollo Scientific
CAS Number:
Specifications
VE:
5 µG
Protein/Peptid-Typ:
Rekombinant
Spezies:
Human
Protein/Peptid-Name:
STAT3
Specifications
About this item
Product Details
Product Details