
4-{[6-(Trifluoromethyl)pyridin-2-yl]oxy}aniline
Catalog # APOSPC6080-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Packung:Bottle
- VE:1 g
- Seq:3047
Specifications
About this item
:
:254.21 g/mol
Storage Temperature:Raumtemperatur
UN:2811
ADR:6.1,III
:GHS06
CAS Number:1086378-51-9
Formula:C₁₂H₉F₃N₂O
MDL Number:MFCD10703507