Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
2-Fluor-4-nitrobenzonitril

2-Fluor-4-nitrobenzonitril

Supplier:   Apollo Scientific
2-Fluor-4-nitrobenzonitril
Supplier:  Apollo Scientific
Certificates
2-Fluor-4-nitrobenzonitril

5 Options Available Below

Product Details & Documents
  • CAS Number:34667-88-4
  • Formula:C₇H₃FN₂O₂
  • MDL Number:MFCD04038715
  • Molecular Weight:166.11 g/mol
  • Storage Temperature:Raumtemperatur
Product Family Options

Product Information

  • Packung
    VE
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - SFr. 26.00
    SFr. 26.00
    1 * 1 g
       SFr. 26.00

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 29.50
    SFr. 29.50
    1 * 5 g
       SFr. 29.50

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 1’840.00
    SFr. 1’840.00
    1 * 500 g
       SFr. 1’840.00

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - SFr. 376.00
    SFr. 376.00
    1 * 100 g
       SFr. 376.00

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - SFr. 101.00
    SFr. 101.00
    1 * 25 g
       SFr. 101.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.