Order Entry
Switzerland
ContactUsLinkComponent
2-Chloro-4-iodo-6-(trifluoromethyl)pyridine 98%
2-Chloro-4-iodo-6-(trifluoromethyl)pyridine 98%
Catalog # APOSPC48602-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-4-iodo-6-(trifluoromethyl)pyridine 98%
Catalog # APOSPC48602-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Packung:
    Bottle
  • VE:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :307.44 g/mol
Melting Pt:93…95 °C
Storage Temperature:Raumtemperatur
CAS Number:205444-22-0
Formula:C₆H₂ClF₃IN
MDL Number:MFCD10699394