
2,6-Difluor-3-methoxyphenylboronsäure
Catalog # APOSPC10334-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Packung:Bottle
- VE:250 mg
- Seq:2590
Specifications
About this item
:
Melting Pt:112…114 °C
:187.94 g/mol
:GHS07
Storage Temperature:Raumtemperatur
CAS Number:870779-02-5
Formula:C₇H₇BF₂O₃
MDL Number:MFCD06657880