
2'-Brom-4'-cyanacetanilid
Catalog # APOSOR5485-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Packung:Bottle
- VE:100 mg
- Seq:2440
Specifications
About this item
:
Storage Temperature:Raumtemperatur
UN:3439
ADR:6.1,III
:GHS07
CAS Number:656834-86-5
Formula:C₉H₇BrN₂O
MDL Number:MFCD03094675