Order Entry
Switzerland
ContactUsLinkComponent
Irritant
2-Cyan-5-methoxybenzoesäure
Catalog # APOSOR21216-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Cyan-5-methoxybenzoesäure
Catalog # APOSOR21216-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Packung:
    Bottle
  • VE:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :177.16 g/mol
 :GHS07
Storage Temperature:Raumtemperatur
CAS Number:179028-65-0
Formula:C₉H₇NO₃
MDL Number:MFCD00116264