Tantal(V)ethanolat 99.999%
Catalog # APOSIN3412-10G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Packung:Bottle
- VE:10 g
- Seq:3236
Specifications
About this item
:
:406.25 g/mol
Boiling Pt:145 °C/0.1mmHg
Melting Pt:21 °C
Density:1.566 g/cm³ (25 °C)
Flash Pt:30 °C
Storage Temperature:Raumtemperatur
EINECS:228-010-2
UN:2920
ADR:8+3,II
:GHS02
:GHS05
CAS Number:6074-84-6
Formula:C₁₀H₂₅O₅Ta
MDL Number:MFCD00049785