Order Entry
Switzerland
ContactUsLinkComponent
(3-Bromobenzylidene)malononitrile
(3-Bromobenzylidene)malononitrile
Catalog # APOSOR111249-1G
Supplier:  Apollo Scientific
CAS Number:  
(3-Bromobenzylidene)malononitrile
Catalog # APOSOR111249-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Packung:
    Bottle
  • VE:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :233.064 g/mol
Storage Temperature:Raumtemperatur
CAS Number:2972-74-9
Formula:C₁₀H₅BrN₂
MDL Number:MFCD00456782