Alafosfalin ≥97%
Anbieter: Apollo Scientific
Direkt zu
Über diesen Artikel
Alaphosphin is an Alanine antagonist and can also be used in Murein biosynthesis.
Synonyme: L-Alanyl-L-1-aminoethylphosphonic acid
- CAS Nummer:60668-24-8
- Formel:C₅H₁₃N₂O₄P
- MDL-Nummer:MFCD01569244
- Molekulargewicht:196.141 g/mol
- Lagertemperatur:Kühlschrank
- EINECS:262-362-8
3 Varianten
Product Details & Documents
Alaphosphin is an Alanine antagonist and can also be used in Murein biosynthesis.
Synonyme: L-Alanyl-L-1-aminoethylphosphonic acid
- CAS Nummer:60668-24-8
- Formel:C₅H₁₃N₂O₄P
- MDL-Nummer:MFCD01569244
- Molekulargewicht:196.141 g/mol
- Lagertemperatur:Kühlschrank
- EINECS:262-362-8