
3,4,5,6-Tetrabromophenolsulphonephthalein 95% (Farbstoffgehalt), Sigma-Aldrich®
Catalog # 227358-5G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- VE:5 g
- Seq:3173
Specifications
About this item
:
:669.96 g/mol
:GHS07
Storage Temperature:Raumtemperatur
CAS Number:77172-72-6
Formula:C₁₉HBr₄O₅S
MDL Number:MFCD00009744