Wipes for general applications, WYPALL* X80 Plus
Catalog # KIMB19139
Anbieter: KIMBERLY CLARK
Technische Daten
- Verpackt:8 Bags with 30 Sheets
- Farbe:Blau
- L×B:350×340 mm
- VE:240
Technische Daten
Über diesen Artikel
The WypAll® X80 Plus Colour-Coded Range has been specially designed for tough cleaning tasks in food processing environments.
- Colour-coded to help avoid cross-contamination
- Wrapped in a splash-proof, water-repellent polybag
- Supplied as quarter-fold cloths in poly packs to provide protection from contamination
- Delivers single sheet dispensing, to control usage, increase hygiene and reduce waste
WypAll® X80 Plus Cloths have been designed with powerpocket embossing for improved cleaning and debris pick up. These cloths are ideal for wet or dry cleaning where a strong and bulky cloth is required and enhance regulatory hygiene practices and compliance.
Advanced Hydroknit technology and powerpocket embossing that is highly absorbent, strong and durable (wet or dry), resists most solvents, reduces residue, allows fast cleaning and helps reduce costs. The embossing offers a great advantage for debris pick-up and gentle scrubbing.
Certifications: Food contact certified.
Delivery: 8 packs×30 quarter fold cloths.