ContactUsLinkComponent
Over-caps, 45 mm, black phenolic, DURAN®
Katalognr. 215-4283
Anbieter:  DWK Life Sciences
undefined
Over-caps, 45 mm, black phenolic, DURAN®
Katalognr. 215-4283
Anbieter:  DWK Life Sciences
Technische Daten
  • Bezeichnung:
    Überkappe, 45 mm schwarzes Phenolharz
  • Material:
    Phenolharz
  • VE:
    10
Technische Daten
Über diesen Artikel
The 45 mm phenolic over-cap screws down over the open-top aluminium GL 45 cap covering and protecting the septum or stopper.
  • Made from high-quality phenol formaldehyde resin (PF)
  • High quality components
  • Recommended usable temperature is 150 °C
The phenolic cap protects the septum or stopper from damage and contamination during storage or shipping. Phenolic caps provide a wide range of chemical compatibility and temperature tolerance.