TCLP Standard 1 for ICP
Catalog # HONCE2160-100ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Matrix:2% HNO₃
- Packung:Plastic bottle
- VE:100 mL
- Bezeichnung:TCLP Standard 1
Specifications
About this item
Certified Reference Material (CRM).
500 mg/L Ba, 100 mg/L Hg, 25 mg/L each of Ag, As, Cr, Pb, 5 mg/L each of Cd, Se in HNO₃ 2%