Primary Interferents Interferents A for ICP
HONCE2114-100ML
:
- Matrix:5% HNO₃
- Packung:Plastic bottle
- VE:100 mL
- Bezeichnung:Primary Interferents Interferents A
Certified Reference Material (CRM).
5000 mg/L each of Al, Ca, Mg, 2000 mg/L Fe in HNO₃ 5%