Instrument Calibration standard 2 for ICP and HGA elements
Catalog # HONCE2104-100ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Matrix:5% HNO₃
- Packung:Plastic bottle
- VE:100 mL
- Bezeichnung:Instrument Calibration Standard 2
Specifications
About this item
Certified Reference Material (CRM).
400 mg/L Ni, 200 mg/L Zn, 150 mg/L Mn, 100 mg/L each of Ag, Cr in HNO₃ 5%