di-Natriumhydrogenphosphat
Catalog # EURLEMR305500
Supplier: EUROCLONE
CAS Number:
Specifications
- VE:1 KIT
- Seq:0075
Specifications
About this item
:
:141.96 g/mol
Merck Index:12,08805
Melting Pt:250 °C
Density:1.53 g/cm³ (20 °C)
Storage Temperature:Raumtemperatur
EINECS:231-448-7
REACH:01-2119489797-11
CAS Number:7558-79-4
Formula:Na₂HPO₄
MDL Number:MFCD00003496