Tetrapropylammoniumhydroxid 0.1 mol/l (0.1 N) in wässriger Lösung
Catalog # DION35363
Supplier: DIONEX
CAS Number:
Specifications
- Packung:Glass bottle
- VE:500 mL
- Seq:2725
Specifications
About this item
:
:203.37 g/mol
Storage Temperature:Kühlschrank
EINECS:224-800-6
CAS Number:4499-86-9
Formula:C₁₂H₂₉NO
MDL Number:MFCD00009360