Folin-Ciocalteus Reagenz für die Phenolbestimmung
Catalog # 31360.264
Supplier: VWR Chemicals
CAS Number:
Specifications
- Packung:Glasflasche
- VE:500 mL
- Seq:2725
Specifications
Specification Test Results
- Identification:Passes test
- Sensitivity to phenol:Passes test
Specification Test Results
About this item
:
Density:1.22 g/cm³ (20 °C)
UN:3264
ADR:8,III
:GHS07
MDL Number:MFCD00132625